missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Anhydrous (USP/FCC/EP), Fisher Chemical™

Catalog No. p-2363482
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S379212 2.5 kg Poly Bottle
S37912 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S379212 Supplier Thermo Fisher Scientific Supplier No. S379212
Only null left

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
pH 8.7 to 9.3
Physical Form Powder/Solid
Quantity 2.5 kg
Formula Weight 141.96
Percent Purity 98.0 to 101.0% (EP)
Grade EP/FCC/USP
Identification Pass Test (USP/FCC/EP), Pass Test (EP), Pass Test (FCC), Pass Test (USP)
Loss on Drying 1.0% max. (EP), 5.0% max. (FCC), 5.0% max. (USP), 5.0% max. (USP/FCC), 1.0% max. (EP)
Packaging Poly Bottle
Chemical Name or Material Sodium Phosphate Dibasic Anhydrous
Show More Show Less