missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Dibasic, Anhydrous, U.S.P. - F.C.C., J.T. Baker™

Catalog No. p-5390941
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Wide Mouth Amber Glass Bottle
Catalog No. Quantity Packaging
02004211 500 g Wide Mouth Amber Glass Bottle
02004223 1 kg Wide Mouth Amber Glass Bottle
2 options

Catalog No. 02004211

Supplier: Avantor 382701

Only null left

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium;hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 500 g
Formula Weight 141.96
Grade FCC/USP
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.07kg
Chemical Name or Material Sodium Phosphate, Dibasic, Anhydrous