missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Dibasic, Anhydrous, Powder, Ultrapure Bioreagent, J.T. Baker™

Catalog No. p-5390940
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
02004198 500 g Poly Bottle
02004199 2.5 kg Poly Bottle
2 options

Catalog No. 02004198

Supplier: Avantor 406201

Only null left

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology applications

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium;hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Powder
Quantity 500 g
Formula Weight 141.96
Grade Ultrapure Bioreagent
Packaging Poly Bottle
Specific Gravity 1L = 2.07kg
Chemical Name or Material Sodium Phosphate, Dibasic, Anhydrous