missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate, Dibasic, Anhydrous, Powder, Ultrapure Bioreagent, J.T. Baker™

Catalog No. 02004199
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004199 2.5 kg Poly Bottle
02004198 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02004199 Supplier Avantor Supplier No. 406205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology applications

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium;hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Powder
Quantity 2.5 kg
Formula Weight 141.96
Grade Ultrapure Bioreagent
Packaging Poly Bottle
Specific Gravity 1L = 2.07kg
Chemical Name or Material Sodium Phosphate, Dibasic, Anhydrous