missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5383217
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
02004224 500 g Poly Bottle
02004225 2.5 kg Poly Bottle
2 options

Catalog No. 02004224

Supplier: Avantor 382801

Only null left

High quality chemicals for laboratory and specialized industrial use

    Suitable for Buffer Solutions

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 500 g
IUPAC Name disodium;hydrogen phosphate
Formula Weight 141.96
Grade Meets ACS reagent chemical requirements
Packaging Poly Bottle
Specific Gravity 1L = 2.07kg
Chemical Name or Material Sodium Phosphate Dibasic, Anhydrous