missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Dibasic, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Description
- Suitable for Buffer Solutions
Specifications
Specifications
| Quantity | 2.5 kg |
| IUPAC Name | disodium hydrogen phosphate |
| Formula Weight | 141.96 |
| Grade | Meets ACS reagent chemical requirements |
| Packaging | Poly bottle |
| Specific Gravity | 1L = 2.07kg |
| Chemical Name or Material | Sodium Phosphate Dibasic, Anhydrous |