missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Sodium Phosphate, Dibasic, ACS Reagent Grade, Calbiochem™,
Supplier: MilliporeSigma™ 5675501KG
| Quantity | 1 kg |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| HNa2O4P | |
| MFCD00003496 | |
| Na2HPO4 | |
| CHEBI:34683 | |
| [Na+].[Na+].OP([O-])([O-])=O |
Specifications
| White | |
| 1 kg | |
| HNa2O4P | |
| Na2HPO4 | |
| [Na+].[Na+].OP([O-])([O-])=O | |
| 141.96 | |
| CHEBI:34683 |
| 7558-79-4 | |
| Glass Bottle | |
| MFCD00003496 | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| disodium hydrogen phosphate | |
| 24203 | |
| ACS Reagent |