missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Dibasic, ACS Grade, ≥ 99.0%, LabChem™
Specifications
Specifications
| Color | White |
| CAS Max % | 100 |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Linear Formula | Na2HPO4 |
| Solubility Information | Soluble in water |
| Formula Weight | 141.96 |
| Percent Purity | ≥99.0% |
| Grade | ACS |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes eye irritation. |
| Precautionary Statement | GHS P Statement Wash exposed skin thoroughly after handling. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| EINECSNumber | 231-448-7 |
| Recommended Storage | Room Temperature |