missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Dibasic, 7-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$3,472.89 - $3,472.89
Chemical Identifiers
| CAS | 7782-85-6 |
|---|---|
| Molecular Formula | H15Na2O11P |
| Molecular Weight (g/mol) | 268.06 |
| MDL Number | MFCD00149180 |
| InChI Key | LVXHNCUCBXIIPE-UHFFFAOYSA-L |
| Synonym | sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate |
| PubChem CID | 6096963 |
| IUPAC Name | disodium phosphoric acid heptahydrate |
| SMILES | O.[Na+].[Na+].OP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
02004221
|
Avantor
382401 |
500 g | Poly Bottle |
N/A
|
|
||||
|
02004222
|
Avantor
382405 |
2.5 kg | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7782-85-6 | |
| 268.06 | |
| LVXHNCUCBXIIPE-UHFFFAOYSA-L | |
| 6096963 | |
| O.[Na+].[Na+].OP([O-])([O-])=O |
| H15Na2O11P | |
| MFCD00149180 | |
| sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
| disodium phosphoric acid heptahydrate |
Specifications
| 7782-85-6 | |
| 500 g | |
| MFCD00149180 | |
| LVXHNCUCBXIIPE-UHFFFAOYSA-L | |
| disodium phosphoric acid heptahydrate | |
| 6096963 | |
| BAKER ANALYZED™ ACS Reagent | |
| 1L = 1.70kg |
| Crystals | |
| H15Na2O11P | |
| sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
| O.[Na+].[Na+].OP([O-])([O-])=O | |
| 268.06 | |
| 268.07 | |
| Poly Bottle | |
| Sodium Phosphate, Dibasic, 7-Hydrate |