missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Dibasic, 7-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Specifications
Specifications
| Physical Form | Crystals |
| Quantity | 2.5 kg |
| Formula Weight | 268.07 |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 1.70kg |
| Chemical Name or Material | Sodium Phosphate, Dibasic, 7-Hydrate |