missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium phosphate, 0.2M buffer soln., pH 8.5

$263.03 - $263.03

Specifications

Chemical Name or Material Sodium phosphate buffer soln.
Concentration 0.2 M
pH 8.5
Physical Form Liquid
MDL Number MFCD00146206
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61372K2
View Documents
Thermo Scientific Chemicals
J61372K2
1 L
Each for $263.03
Only null left
 
Description

Description

Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

Solubility
Soluble in water (121 mg/ml at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong acids.
Specifications

Specifications

Sodium phosphate buffer soln.
8.5
MFCD00146206
0.2 M
Liquid
Soluble in water (121mg/ml at 20°C).
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only