missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium phosphate, 0.2M buffer soln., pH 8.0

Catalog No. AAJ62733AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62733AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62733AP Supplier Thermo Scientific Chemicals Supplier No. J62733AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphate buffer is a multi-purpose buffer used for many molecular biology procedures. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphate buffer is a multi-purpose buffer used for many molecular biology procedures. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

Solubility
Fully miscible in water.

Notes
Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications

Chemical Name or Material Sodium phosphate buffer soln.
Concentration 0.2 M
pH 8.0
Physical Form Liquid
Quantity 500 mL
MDL Number MFCD00146206
Solubility Information Fully miscible in water.

RUO – Research Use Only