missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium phosphate, 0.2M buffer soln., pH 7.4
Description
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately. It is also used in the preparation of mobile phase in high performance liquid chromatography (HPLC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately. It is also used in the preparation of mobile phase in high performance liquid chromatography (HPLC).
Solubility
Miscible with water. Immiscible with alcohol.
Notes
Incompatible with strong acids.
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately. It is also used in the preparation of mobile phase in high performance liquid chromatography (HPLC).
Solubility
Miscible with water. Immiscible with alcohol.
Notes
Incompatible with strong acids.
Specifications
Specifications
| Chemical Name or Material | Sodium phosphate buffer soln. |
| Concentration | 0.2 M |
| pH | 7.4 |
| Physical Form | Liquid |
| Quantity | 1 L |
| MDL Number | MFCD00146206 |
| Solubility Information | Miscible with water. Immiscible with alcohol. |
RUO – Research Use Only