missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phenylpyruvate monohydrate, 98%
Description
Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Linear Formula | C6H5CH2COCO2Na·H2O |
| Quantity | 25 g |
| Beilstein | 4770978 |
| Solubility Information | Soluble in water. |
| Formula Weight | 204.16 (186.14 Anhydrous) |
| Percent Purity | 98% |
| Chemical Name or Material | Sodium phenylpyruvate monohydrate |
Safety and Handling
| EINECSNumber | 204-053-2 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only