missing translation for 'onlineSavingsMsg'
Learn More

Sodium phenylpyruvate monohydrate, 98%

Catalog No. aaa1402706
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
50 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1402706 5 g
AAA1402703 1 g
AAA1402714 25 g
AAA1402718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1402706 Supplier Thermo Scientific Chemicals Supplier No. A1402706
Only null left

CAS: 122049-54-1 | C9H7O3 | 163.15 g/mol

Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 122049-54-1
Molecular Formula C9H7O3
Molecular Weight (g/mol) 163.15
MDL Number MFCD00149109
InChI Key BTNMPGBKDVTSJY-UHFFFAOYSA-M
Synonym sodium phenylpyruvate monohydrate, sodium 2-oxo-3-phenylpropanoate hydrate, phenylpyruvic acid sodium salt monohydrate, phenylpyruvic acid, sodium salt, sodium hydrate ?-phenylpyruvate, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, acmc-2099lh, phenylpyruvic acid sodium hydrate, beta-phenylpyruvic acid, sodium salt
PubChem CID 23666336
IUPAC Name sodium;2-oxo-3-phenylpropanoate;hydrate
SMILES [O-]C(=O)C(=O)CC1=CC=CC=C1

Specifications

Melting Point >300°C
Linear Formula C6H5CH2COCO2Na·H2O
Quantity 5 g
Beilstein 4770978
Solubility Information Soluble in water.
Formula Weight 204.16 (186.14 Anhydrous)
Percent Purity 98%
Chemical Name or Material Sodium phenylpyruvate monohydrate
EINECSNumber 204-053-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only