missing translation for 'onlineSavingsMsg'
Learn More

Sodium phenylpyruvate, 98%

Catalog No. AAH5676706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5676706 5 g
AAH5676714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5676706 Supplier Thermo Scientific Chemicals Supplier No. H5676706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 114-76-1 | C9H7NaO3 | 186.142 g/mol

Phenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.

Solubility
Soluble in water

Notes
Store at 2-8°C. Avoin oxidizing agents.

Chemical Identifiers

CAS 114-76-1
Molecular Formula C9H7NaO3
Molecular Weight (g/mol) 186.142
MDL Number MFCD00002590
InChI Key MQGYVGKMCRDEAF-UHFFFAOYSA-M
Synonym sodium phenylpyruvate, phenylpyruvic acid sodium salt, sodium 2-oxo-3-phenylpropanoate, unii-7528cw33ww, sodium ?-phenylpyruvate, phenylpyruvic acid, sodium salt, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, 2-oxo-3-phenylpropionic acid, 3-phenyl-2-oxopropanoic acid, sodium salt
PubChem CID 23667645
IUPAC Name sodium;2-oxo-3-phenylpropanoate
SMILES C1=CC=C(C=C1)CC(=O)C(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 4770978
Solubility Information Soluble in water
Formula Weight 186.14
Percent Purity 98%
Chemical Name or Material Sodium phenylpyruvate
EINECSNumber 204-053-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only