missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phenoxyacetate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 264°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 174.13 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Sodium Phenoxyacetate |
Safety and Handling
| RTECSNumber | AJ2400000 |
| TSCA | Yes |