accessibility menu, dialog, popup

UserName

NaOOO

CAS RN 3598-16-1

IUPAC Name: sodium 2-phenoxyacetate
Synonyms: sodium phenoxyacetate
Molecular Weight (g/mol): 174.13
Molecular Formula: C8H7NaO3
InChi Key: WPTJBFNYRRZIDZ-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)COC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results