Learn More
Sodium Phenate Solution, For Nitrogen (Ammonia), Nitrogen (Organic), LabChem™
Supplier: LabChem LC247702
Specifications
| Sodium Phenate Solution | |
| For Nitrogen (Ammonia), Nitrogen (Organic) | |
| 88.4,8.4,3.2 | |
| UN1760 | |
| Soluble in water | |
| Passes Test | |
| Carbon monoxide; Carbon dioxide | |
| Liquid |
| Amber to Colorless | |
| 7732-18-5,108-95-2,1310-73-2 | |
| C6H5O8 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| 94.11 | |
| Poly Bottle | |
| 1 L |
Chemical Identifiers
| 7732-18-5 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Suspected of causing genetic defects.
May cause damage to organs (kidneys, liver) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Use only outdoors or in a well-ventilated area.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature