missing translation for 'onlineSavingsMsg'
Learn More

Sodium Persulfate, 1.00 M, Ricca Chemical

Catalog No. R75150001BV
Change view
Click to view available options
Quantity:
1 L
10 L
Packaging:
Cubitainer
Vented Opaque Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R75150001BV 1 L Vented Opaque Poly Bottle
R751500010F 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R75150001BV Supplier Ricca Chemical Company Supplier No. R75150001BV

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7775-27-1 , 7732-18-5
Molecular Formula Na2O8S2
Molecular Weight (g/mol) 238.092
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
PubChem CID 62655
IUPAC Name disodium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 1 L
Assay Percent Range 1.000 ± 0.005 M at 20°C
Odor Odorless
Packaging Vented Opaque Poly Bottle
Chemical Name or Material Sodium Persulfate, 1.00 Molar