missing translation for 'onlineSavingsMsg'
Learn More

Sodium Peroxide, Technical, 85%, Spectrum™ Chemical

Catalog No. p-7166345
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613777 25 g Steel Can
18613774 125 g Steel Can
18613775 500 g Steel Can
18613776 2.5 kg Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613777 Supplier Spectrum Chemical Mfg Cor Supplier No. S139225GM
Only null left

Chemical Identifiers

CAS 1313-60-6

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 85%
Grade Technical
Packaging Steel Can

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.