missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Peroxide, Reagent grade, Honeywell™
Reagent Grade, granular, +140Â mesh particle size, 97%
$1409.26 - $1409.26
Chemical Identifiers
| CAS | 1313-60-6 |
|---|---|
| Molecular Formula | Na2O2 |
| Molecular Weight (g/mol) | 77.978 |
| MDL Number | MFCD00003497 |
| InChI Key | PFUVRDFDKPNGAV-UHFFFAOYSA-N |
| Synonym | sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide |
| PubChem CID | 14803 |
| IUPAC Name | disodium;peroxide |
| SMILES | [O-][O-].[Na+].[Na+] |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
| 1313-60-6 | |
| 77.978 | |
| PFUVRDFDKPNGAV-UHFFFAOYSA-N | |
| 14803 | |
| [O-][O-].[Na+].[Na+] |
| Na2O2 | |
| MFCD00003497 | |
| sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide | |
| disodium;peroxide |
Specifications
| 1313-60-6 | |
| 500 g | |
| MFCD00003497 | |
| sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide | |
| [O-][O-].[Na+].[Na+] | |
| 77.978 | |
| 77.98g/mol | |
| Reagent |
| Granules | |
| Na2O2 | |
| 1504 | |
| PFUVRDFDKPNGAV-UHFFFAOYSA-N | |
| disodium;peroxide | |
| 14803 | |
| 97% | |
| Sodium Peroxide |
Safety and Handling
P220-P280-P305 + P351 + P338-P310
H271-H314
EINECSNumber : 215-209-4