missing translation for 'onlineSavingsMsg'
Learn More

Sodium Peroxide, puriss. p.a. ACS Reagent, Solstice

Catalog No. 6001537
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001537 100 g
6001539 500 g
6001538 1 kg
60045956 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6001537 Supplier Solstice Supplier No. 31445100G

Puriss. p.a., ACS Reagent, Reag. ISO,≥95%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.978
MDL Number MFCD00003497
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
Synonym sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
PubChem CID 14803
IUPAC Name disodium;peroxide
SMILES [O-][O-].[Na+].[Na+]

Specifications

Quantity 100 g
UN Number 1504
Formula Weight 77.98g/mol
Percent Purity ≥95%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium Peroxide, Reag. ISO
Hazard Statement P220-P280-P305 + P351 + P338-P310
Precautionary Statement H271-H314
EINECSNumber 215-209-4