Learn More
Sodium peroxide, ACS, 93% min
For Use As An Oxidizing Agent, a Bleaching Agent, Deodorant | CAS: 1313-60-6 | Na2O2 | 77.978 g/mol
Supplier: Thermo Scientific Chemicals 03338422
Description
As an oxidizing agent, a bleaching agent, deodorant, pharmaceuticals and dyes, and as an analytical reagent.Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium peroxide | |
| For analysis ACS | |
| White to Yellow | |
| 100 g | |
| Na2O2 | |
| sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide | |
| PFUVRDFDKPNGAV-UHFFFAOYSA-N | |
| disodium;peroxide | |
| 14803 | |
| ACS | |
| White to pale yellow granules |
| 460°C (decomposition) | |
| 1313-60-6 | |
| Granules | |
| 93% min. | |
| MFCD00003497 | |
| Soluble in water,forming NaOH and H2O2. Soluble in acid. Insoluble in alkali. | |
| [O-][O-].[Na+].[Na+] | |
| 77.978 | |
| 77.98 | |
| Air sensitive; Hygroscopic | |
| 2.805 g/cm3 at 20°C |
Chemical Identifiers
| 1313-60-6 | |
| 77.978 | |
| PFUVRDFDKPNGAV-UHFFFAOYSA-N | |
| 14803 | |
| [O-][O-].[Na+].[Na+] |
| Na2O2 | |
| MFCD00003497 | |
| sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide | |
| disodium;peroxide |
Safety and Handling
P210-P220-P221-P234-P260-P264b-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P390-P501c
H271-H290-H314-H335
DOTInformation : DOT Class: 5.1; Packing Group: I
EINECSNumber : 215-209-4
RTECSNumber : WD3450000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only