missing translation for 'onlineSavingsMsg'
Learn More

Sodium peroxide, ACS, 93% min

Catalog No. AA3338409
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3338409 10 g
AA3338422 100 g
AA3338436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3338409 Supplier Thermo Scientific Chemicals Supplier No. 03338409

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Use As An Oxidizing Agent, a Bleaching Agent, Deodorant ∣ CAS: 1313-60-6 ∣ Na2O2 ∣ 77.978 g/mol

As an oxidizing agent, a bleaching agent, deodorant, pharmaceuticals and dyes, and as an analytical reagent.Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.978
MDL Number MFCD00003497
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
Synonym sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
PubChem CID 14803
IUPAC Name disodium;peroxide
SMILES [O-][O-].[Na+].[Na+]

Specifications

Melting Point 460°C (decomposition)
Color White to Yellow
Physical Form Granules
Quantity 10 g
Sensitivity Air sensitive; Hygroscopic
Appearance White to pale yellow granules
Assay Percent Range 93% min.
Density 2.805 g/cm3 at 20°C
Solubility Information Soluble in water,forming NaOH and H2O2. Soluble in acid. Insoluble in alkali.
Formula Weight 77.98
Percent Purity ≥93%
Grade ACS
Chemical Name or Material Sodium peroxide, For analysis ACS
Show More Show Less
Hazard Category H271-H290-H314-H335
Precautionary Statement P210-P220-P221-P234-P260-P264b-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P390-P501c
DOTInformation DOT Class: 5.1; Packing Group: I
EINECSNumber 215-209-4
RTECSNumber WD3450000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only