missing translation for 'onlineSavingsMsg'
Learn More

Sodium peroxide, 95%

CAS: 1313-60-6 | Na2O2 | 77.978 g/mol

Supplier:  Thermo Scientific Chemicals L133060B

 View more versions of this product

Catalog No. AAL133060B


Only null left
Explore available promotions

Description

Description

Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.

Solubility
Soluble in acid. Insoluble in alkali.

Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. It reacts with water violently to give sodium hydroxide and hydrogen peroxide. Incompatible with strong reducing agents, organic materials, alcohols and powdered metals.
Specifications

Specifications

Sodium peroxide
1313-60-6
Na2O2
UN1504
sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
PFUVRDFDKPNGAV-UHFFFAOYSA-N
disodium;peroxide
14803
95%
2.805
460°C (decomposition)
1000 g
MFCD00003497
14,8655
Soluble in acid. Insoluble in alkali.
[O-][O-].[Na+].[Na+]
77.978
77.98
Air sensitive; Hygroscopic

Chemical Identifiers

1313-60-6
77.978
PFUVRDFDKPNGAV-UHFFFAOYSA-N
14803
[O-][O-].[Na+].[Na+]
Na2O2
MFCD00003497
sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
disodium;peroxide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H271-H314
May cause fire or explosion; strong oxidizer.
Causes severe skin burns and eye damage.

P210-P220-P221-P234-P260-P264b-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P390-P501c

H271-H290-H314-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: SODIUM PEROXIDE

EINECSNumber : 215-209-4

RTECSNumber : WD3450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only