missing translation for 'onlineSavingsMsg'
Learn More

Sodium periodate, 99.8+%, ACS reagent

Catalog No. AC419615000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419615000 500 g Plastic bottle
AC419610050 5 g Glass Bottle
AC419611000 100 g Plastic bottle
AC419610025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC419615000 Supplier Thermo Scientific Chemicals Supplier No. 419615000

CAS: 7790-28-5 | INaO4 | 213.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
MDL Number MFCD00003534
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
PubChem CID 23667635
ChEBI CHEBI:75226
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Melting Point 300.0°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99.8 to 100.3% (on dried basis) (Iodometry)
Linear Formula NaIO4
Fieser 01,809; 02,311; 04,373; 05,507; 06,455; 11,492; 13,145; 15,294
Merck Index 15,8772
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: 389g/L (52°C), soluble in h2so4, hno3, acetic acids, insoluble in most common organic solvents
IUPAC Name sodium;periodate
Formula Weight 213.89
Percent Purity ≥99.8%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium periodate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause fire or explosion; strong oxidizer.
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure if swallowed.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
EINECSNumber 232-197-6

RUO – Research Use Only