missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate, Anhydrous, Reagent, ACS, 98-102%, Spectrum™ Chemical

S1377, 7601-89-0, NaClO4

Supplier:  Spectrum Chemical Mfg Cor S1377500GM

 View more versions of this product

Catalog No. 18606688


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Perchlorate, Anhydrous, Reagent, ACS
Specifications

Specifications

7601-89-0
6.0 to 8.0
ClNaO4
[Na+].[O-][Cl](=O)(=O)=O
122.44
ACS
1
500 g
BAZAXWOYCMUHIX-UHFFFAOYSA-M
sodium perchlorate
98 to 102%
Amber Glass Bottle

Chemical Identifiers

7601-89-0
122.44
sodium perchlorate
ClNaO4
BAZAXWOYCMUHIX-UHFFFAOYSA-M
[Na+].[O-][Cl](=O)(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions