missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate, Anhydrous, Reagent, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18606686
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606686 25 g Amber Glass Bottle
18606687 125 g Amber Glass Bottle
18606688 500 g Amber Glass Bottle
18606689 2.5 kg Amber Glass Bottle
18606690 12 kg Poly Pail
18606691 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18606686 Supplier Spectrum Chemical Mfg Cor Supplier No. S137725GM
Only null left

S1377, 7601-89-0, NaClO4

Spectrum™ Chemical Sodium Perchlorate, Anhydrous, Reagent, ACS

Chemical Identifiers

CAS 7601-89-0
Molecular Formula ClNaO4
Molecular Weight (g/mol) 122.44
InChI Key BAZAXWOYCMUHIX-UHFFFAOYSA-M
IUPAC Name sodium perchlorate
SMILES [Na+].[O-][Cl](=O)(=O)=O

Specifications

pH 6.0 to 8.0
CAS Max % 1
Quantity 25 g
Percent Purity 98 to 102%
Grade ACS
Packaging Amber Glass Bottle