missing translation for 'onlineSavingsMsg'
Learn More

Sodium perchlorate, anhydrous, ACS, 98.0-102.0%

Catalog No. p-4407924
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1162322 100 g
AA1162336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1162322 Supplier Thermo Scientific Chemicals Supplier No. 01162322
Only null left

CAS: 7601-89-0 | ClNaO4 | 122.436 g/mol

Sodium perchlorate is used in chemical analysis and in explosive materials, fireworks, and rocket motors. It is the precursor for the preparation of other perchlorate salts. It is also employed as a chaotrophic agent in DNA extraction and hybridization reactions in molecular biology. It prevents the binding of phenolic compounds to RNA during extraction. In addition, it is used in batteries, plastics and in rubber products. It is also used as a nonreactive electrolyte. Further, it is an eluent component used for analysis of biological samples by liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7601-89-0
Molecular Formula ClNaO4
Molecular Weight (g/mol) 122.436
MDL Number MFCD00003480
InChI Key BAZAXWOYCMUHIX-UHFFFAOYSA-M
Synonym sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
PubChem CID 522606
IUPAC Name sodium;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Na+]

Specifications

Melting Point 482°C (decomposition)
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98.0 to 102.0%
Solubility Information Soluble in water and alcohol.
Formula Weight 122.46
Grade ACS Reagent
Sensitivity Hygroscopic
Density 2.50 g/mL
Chemical Name or Material Sodium perchlorate, anhydrous, For analysis ACS
Hazard Category H271-H302-H319-H373
Precautionary Statement P210-P220-P221-P260-P264b-P270-P280-P283-P301+P312-P305+P351+P338-P306+P360-P314-P330-P370+P378q-P371+P380+P375-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 231-511-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only