missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perborate, Monohydrate, Spectrum™ Chemical

Catalog No. 18613757
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613757 500 g Poly Pail
18613756 2.5 kg Poly Pail
18613755 12 kg Poly Pail
18613758 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613757 Supplier Spectrum Chemical Mfg Cor Supplier No. S1589500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10332-33-9

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 500 g
Molecular Weight (g/mol) 99.81
Grade Ungraded
Packaging Poly Pail
Chemical Name or Material Sodium Perborate, Monohydrate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.