missing translation for 'onlineSavingsMsg'
Learn More

Sodium Palmitate 97.0+%, TCI America™

Catalog No. P000725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P000725G 25 g
P0007500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P000725G Supplier TCI America Supplier No. P000725G

Chemical Identifiers

CAS 408-35-5
Molecular Formula C16H31NaO2
Molecular Weight (g/mol) 278.412
MDL Number MFCD00002749
InChI Key GGXKEBACDBNFAF-UHFFFAOYSA-M
Synonym sodium palmitate, palmitic acid sodium salt, hexadecanoic acid, sodium salt, sodium hexadecanoate, palmitic acid, sodium salt, sodium pentadecanecarboxylate, unii-jq43kp6296, hsdb 759, hexadecanoic acid sodium salt, hexadecanoic acid,sodium salt 1:1
PubChem CID 2735111
IUPAC Name sodium;hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 278.41
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Palmitate
EINECSNumber (2)-0611&(7)-0973
RTECSNumber RT4945000
TSCA Yes