accessibility menu, dialog, popup

UserName

NaH3COO

CAS RN 408-35-5

IUPAC Name: sodium hexadecanoate
Synonyms: palmitate, sodium
Molecular Weight (g/mol): 278.41
Molecular Formula: C16H31NaO2
InChi Key: GGXKEBACDBNFAF-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)