missing translation for 'onlineSavingsMsg'
Learn More

Sodium p-Toluenesulfinate Tetrahydrate 98.0+%, TCI America™

Catalog No. T076325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T076325G 25 g
T0763500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T076325G Supplier TCI America Supplier No. T076325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium p-Toluenesulfinate Tetrahydrate 98.0+%

Chemical Identifiers

CAS 868858-48-4
Molecular Formula C7H15NaO6S
Molecular Weight (g/mol) 250.241
MDL Number MFCD00149644
InChI Key MLPRYQZBDONMGJ-UHFFFAOYSA-M
Synonym p-Toluenesulfinic Acid Sodium Salt
PubChem CID 23690860
IUPAC Name sodium;4-methylbenzenesulfinate;tetrahydrate
SMILES CC1=CC=C(C=C1)S(=O)[O-].O.O.O.O.[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 178.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium p-Toluenesulfinate Tetrahydrate
EINECSNumber (3)-1873
RTECSNumber XT4725000
TSCA Yes