accessibility menu, dialog, popup

UserName

CH3SOOH2OH2OH2OH2ONa

CAS RN 868858-48-4

IUPAC Name: sodium 4-methylbenzene-1-sulfinate tetrahydrate
Synonyms: sodium 4-methylbenzenesulfinate tetrahydrate
Molecular Weight (g/mol): 250.24
Molecular Formula: C7H15NaO6S
InChi Key: MLPRYQZBDONMGJ-UHFFFAOYSA-M
SMILES: O.O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)