missing translation for 'onlineSavingsMsg'
Learn More

Sodium p-Styrenesulfonate Hydrate 93.0+%, TCI America™

Catalog No. S0258500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0258500G 500 g
S025825G 25 g
S0258100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S0258500G Supplier TCI America Supplier No. S0258500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123333-94-8
Molecular Formula C8H9NaO4S
Molecular Weight (g/mol) 224.21
MDL Number MFCD03092905
InChI Key AATHLPHPRXGBAI-UHFFFAOYSA-M
Synonym p-Styrenesulfonic Acid Sodium Salt
PubChem CID 23681144
IUPAC Name sodium 4-ethenylbenzene-1-sulfonate hydrate
SMILES O.[Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1

Specifications

Color White
Quantity 500 g
Formula Weight 206.19
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium p-Styrenesulfonate Hydrate
EINECSNumber (3)-1903&(3)-1948
TSCA Yes