missing translation for 'onlineSavingsMsg'
Learn More

Sodium oxalate, Certified by BAM, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111014938
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014938 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014938 Supplier Merck Emd Millipore Supplier No. 7180425G
Only null left

Specifications

CAS 62-76-0
Physical Form Neat
Linear Formula NaOOCCOONa
MDL Number MFCD00012465
Synonym Ethanedioic acid sodium salt; Oxalic acid disodium salt
Type Certified Reference Material
Quantity 25 g
Formula Weight 134.00
Percent Purity ≥99.5%
Grade Certified Reference Material