missing translation for 'onlineSavingsMsg'
Learn More

Sodium oxalate, Acculute Standard Volumetric Solution, Final Concentration 0.1N

Catalog No. p-4782152
Change view
Click to view available options
Quantity:
1 Unit
6 Units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA88626LQ 1 Unit
AA88626LS 6 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA88626LQ Supplier Thermo Scientific Chemicals Supplier No. 088626LQ
Only null left

CAS: 62-76-0 | C2Na2O4 | 134.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
PubChem CID 6125
IUPAC Name disodium;oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Odor Odorless
Physical Form Liquid
Color Colorless
Quantity 1 Unit
Formula Weight 134
Chemical Name or Material Sodium oxalate
Hazard Category H314-H335-H350
Hazard Statement Avoid contact with skin and eyes.
Precautionary Statement P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
EINECSNumber 200-550-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only