missing translation for 'onlineSavingsMsg'
Learn More

Sodium orthovanadate, 99.9% (metals basis)

inhibitor of Protein Tyrosine Phosphatase and Alkaline Phosphate | CAS: 13721-39-6 | Na3O4V | 183.907 g/mol

$168.42 - $2469.66

Chemical Identifiers

CAS 13721-39-6
Molecular Formula Na3O4V
Molecular Weight (g/mol) 183.907
MDL Number MFCD00003511
InChI Key IHIXIJGXTJIKRB-UHFFFAOYSA-N
Synonym sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
PubChem CID 61671
ChEBI CHEBI:35607
IUPAC Name trisodium;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA8110414
View Documents
Thermo Scientific Chemicals
08110414
25 g
Each for $168.42
Only null left
 
AA8110422
View Documents
Thermo Scientific Chemicals
08110422
100 g
Each for $563.14
Only null left
 
AA8110436
View Documents
Thermo Scientific Chemicals
08110436
500 g
Each for $2,469.66
Only null left
 
Description

Description

Inhibitor of protein tyrosine phosphatase and alkaline phosphate Sodium orthovanadate is used in protein analysis in molecular biology and acts as an inhibitor of protein tyrosine phosphatases, alkaline phosphatases and kinases. It is also used to prepare the stop solution in dephosphorylation assay of insulin receptor kinase. It serves as a reagent in the development of matrix Chip that utilizes surface-immobilized antibodies. Further, it is utilized for the preservation of protein tyrosyl phosphorylation state in cells and cell lysates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13721-39-6
183.907
IHIXIJGXTJIKRB-UHFFFAOYSA-N
61671
trisodium;trioxido(oxo)vanadium
Na3O4V
MFCD00003511
sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
CHEBI:35607
[O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

850°C to 866°C
White
25 g
Na3O4V
sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
IHIXIJGXTJIKRB-UHFFFAOYSA-N
trisodium;trioxido(oxo)vanadium
61671
183.94
Sodium orthovanadate
13721-39-6
Granules
99.9% (metals basis)
MFCD00003511
Soluble in water.
[O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]
183.907
CHEBI:35607
Odorless
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 237-287-9

RTECSNumber : YW1120000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only