missing translation for 'onlineSavingsMsg'
Learn More

Sodium orthovanadate, 99.9% (metals basis)

inhibitor of Protein Tyrosine Phosphatase and Alkaline Phosphate | CAS: 13721-39-6 | Na3O4V | 183.907 g/mol

Supplier:  Thermo Scientific Chemicals 08110436

 View more versions of this product

Catalog No. AA8110436


Only null left
Explore available promotions

Description

Description

Inhibitor of protein tyrosine phosphatase and alkaline phosphate Sodium orthovanadate is used in protein analysis in molecular biology and acts as an inhibitor of protein tyrosine phosphatases, alkaline phosphatases and kinases. It is also used to prepare the stop solution in dephosphorylation assay of insulin receptor kinase. It serves as a reagent in the development of matrix Chip that utilizes surface-immobilized antibodies. Further, it is utilized for the preservation of protein tyrosyl phosphorylation state in cells and cell lysates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium orthovanadate
13721-39-6
Granules
99.9% (metals basis)
MFCD00003511
Soluble in water.
[O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]
183.907
CHEBI:35607
Odorless
850°C to 866°C
White
500 g
Na3O4V
sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
IHIXIJGXTJIKRB-UHFFFAOYSA-N
trisodium;trioxido(oxo)vanadium
61671
183.94

Chemical Identifiers

13721-39-6
183.907
IHIXIJGXTJIKRB-UHFFFAOYSA-N
61671
trisodium;trioxido(oxo)vanadium
Na3O4V
MFCD00003511
sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
CHEBI:35607
[O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 237-287-9

RTECSNumber : YW1120000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only