missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Orthovanadate, 96% min., (Powder), MP Biomedicals
Potent alkaline phosphate inhibitor
Manufacturer: Mp Biomedicals 0215966480
Specifications
Sodium Orthovanadate | |
100g | |
Na3O4V | |
IHIXIJGXTJIKRB-UHFFFAOYSA-N | |
trisodium;trioxido(oxo)vanadium | |
61671 | |
183.9 |
13721-39-6 | |
96% min. | |
sodium o-vanadate, sodium orthovanadate, sodium pervanadate, sodium tetraoxovanadate 3-, sodium vanadate ortho, trisodium orthovanadate, trisodium tetraoxovanadate, trisodium vanadate, vanadic acid h3vo4 , sodium salt, vanadic acid, trisodium salt | |
[O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+] | |
183.907 | |
CHEBI:35607 | |
Powder |
Chemical Identifiers
13721-39-6 | |
183.907 | |
sodium o-vanadate, sodium orthovanadate, sodium pervanadate, sodium tetraoxovanadate 3-, sodium vanadate ortho, trisodium orthovanadate, trisodium tetraoxovanadate, trisodium vanadate, vanadic acid h3vo4 , sodium salt, vanadic acid, trisodium salt | |
CHEBI:35607 | |
[O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+] |
Na3O4V | |
IHIXIJGXTJIKRB-UHFFFAOYSA-N | |
61671 | |
trisodium;trioxido(oxo)vanadium |
Safety and Handling
Recommended Storage : Room Temperature