missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Orthovanadate, 96% min., (Powder), MP Biomedicals
Potent alkaline phosphate inhibitor
$389.46 - $389.46
Chemical Identifiers
| CAS | 13721-39-6 |
|---|---|
| Molecular Formula | Na3O4V |
| Molecular Weight (g/mol) | 183.907 |
| InChI Key | IHIXIJGXTJIKRB-UHFFFAOYSA-N |
| Synonym | sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt |
| PubChem CID | 61671 |
| ChEBI | CHEBI:35607 |
| IUPAC Name | trisodium;trioxido(oxo)vanadium |
| SMILES | [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15966480
|
MP Biomedicals Inc
0215966480 |
100 g |
N/A
|
|
|||||
Chemical Identifiers
| 13721-39-6 | |
| 183.907 | |
| sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt | |
| CHEBI:35607 | |
| [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+] |
| Na3O4V | |
| IHIXIJGXTJIKRB-UHFFFAOYSA-N | |
| 61671 | |
| trisodium;trioxido(oxo)vanadium |
Specifications
| 13721-39-6 | |
| 100 g | |
| Na3O4V | |
| IHIXIJGXTJIKRB-UHFFFAOYSA-N | |
| trisodium;trioxido(oxo)vanadium | |
| 61671 | |
| 183.9 |
| Powder | |
| 96% min. | |
| sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt | |
| [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+] | |
| 183.907 | |
| CHEBI:35607 | |
| Sodium Orthovanadate |
Safety and Handling
Recommended Storage : Room Temperature