missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroprusside Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017850
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017850 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017850 Supplier Merck Emd Millipore Supplier No. PHR14231G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. Sodium Nitroprusside is widely used as a drug to manage hypertension, via lowering elevated blood pressure by acting on the arteriolar smooth muscle.

Specifications

CAS 13755-38-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula Na2 [Fe(CN)5 NO] · 2H2 O
MDL Number MFCD00149192
Synonym Sodium nitroferricyanide(III) dihydrate; Nitroprusside sodium; SNP; Sodium nitroprusside; Sodium pentacyanonitrosylferrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 297.95
Grade Certified Reference Material