missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitroferricyanide(Iii) Dihydrate, Honeywell Fluka™
ACS Reagent,≥99%
$1495.05 - $1495.05
Chemical Identifiers
| CAS | 13755-38-9 |
|---|---|
| Molecular Formula | C5H4FeN6Na2O3 |
| Molecular Weight (g/mol) | 297.95 |
| MDL Number | MFCD00149192 |
| InChI Key | JFPDVRGGFBUORC-UHFFFAOYSA-N |
| Synonym | nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o |
| PubChem CID | 11953895 |
| SMILES | O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 13755-38-9 | |
| 297.95 | |
| JFPDVRGGFBUORC-UHFFFAOYSA-N | |
| 11953895 |
| C5H4FeN6Na2O3 | |
| MFCD00149192 | |
| nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o | |
| O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] |
Specifications
| 13755-38-9 | |
| C5H4FeN6Na2O3 | |
| MFCD00149192 | |
| nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o | |
| O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] | |
| 297.95 | |
| 297.95g/mol | |
| ACS Reagent |
| 500 g | |
| Na2[Fe(CN)5NO] · 2H2O | |
| 3288 | |
| JFPDVRGGFBUORC-UHFFFAOYSA-N | |
| disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate | |
| 11953895 | |
| ≥99% | |
| Sodium Nitroferricyanide(Iii) Dihydrate |
Safety and Handling
P301 + P310
H301
EINECSNumber : 238-373-9
RTECSNumber : LJ8925000