missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroferricyanide(Iii) Dihydrate, Solstice

Catalog No. 6001518
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001518 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001518 Supplier Solstice Supplier No. 228710500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Physical Form Crystals
Quantity 500 g
Linear Formula Na2[Fe(CN)5NO] · 2H2O
UN Number 3288
IUPAC Name disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
Formula Weight 297.95g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Sodium Nitroferricyanide(Iii) Dihydrate
Hazard Statement P301 + P310
Precautionary Statement H301
EINECSNumber 238-373-9
RTECSNumber LJ8925000