missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrate, Purified, Reagents

Catalog No. p-200020945
Change view
Click to view available options
Quantity:
100 lb.
2.5 kg
500 g
Catalog No. Quantity
01337000 100 lb.
01336999 2.5 kg
01336998 500 g
3 options

Catalog No. 01337000

Supplier: Labchem Inc C2320500100F5

Only null left

Sodium Nitrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym Nitrate of Soda
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Boiling Point 380°C
Melting Point 306°C
Color Colorless/White
Physical Form Solid
Quantity 100 lb.
Grade Pure
Packaging Fiber Drum
Specific Gravity 2.26
Chemical Name or Material Sodium Nitrate, Purified