missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, meets analytical spec. of FCC, E251, 99.0-100.5%, Honeywell Fluka™

Catalog No. 6001505
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001505 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001505 Supplier Honeywell Inc Supplier No. 134441KG

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Meets analytical specification of FCC, E251, 99.0-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
PubChem CID 24268
ChEBI CHEBI:63005
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Quantity 1 kg
Linear Formula NaNO3
UN Number 1498
Solubility Information Soluble in water 1gm/1.1mL, Soluble in alcohol 1gm/125mL, Soluble in boling water 1gm/0.6mL, Soluble in boiling alcohol 1gm/52mL
IUPAC Name sodium;nitrate
Formula Weight 84.99g/mol
Percent Purity 99 to 100.5%
Chemical Name or Material Sodium Nitrate, Meets analytical specification of FCC
Hazard Statement P210-P220-P221-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H319
EINECSNumber 231-554-3
RTECSNumber WC5600000