missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium nitrate, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 380.0°C |
| Melting Point | 306.0°C |
| Color | Colorless to White |
| pH | 5.5 to 8.3 (5% aq. soln.) |
| Physical Form | Crystalline Powder and/or Crystals |
| Quantity | 5 kg |
| Loss on Drying | 2% max. (105°C) |
| Packaging | Plastic Bottle |
| Linear Formula | NaNO3 |
| Merck Index | 15,8778 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May intensify fire; oxidizer. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. In case of major |
| EINECSNumber | 231-554-3 |
RUO – Research Use Only