missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrate (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical S3433

 View more versions of this product

Catalog No. S343-3



Only null left
Explore available promotions

Description

Description

Specifications

Specifications

380°C
306°C
White
Crystalline Solid
≥99 %
MFCD00011119
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
[Na+].[O-][N+]([O-])=O
84.99
CHEBI:63005
≥99%
Pass Test
Sodium Nitrate
7631-99-4
5.5 to 8
3 kg
NNaO3
1498
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium nitrate
24268
84.99
Certified ACS
Poly Bottle

Chemical Identifiers

7631-99-4
84.99
VWDWKYIASSYTQR-UHFFFAOYSA-N
24268
sodium nitrate
NNaO3
MFCD00011119
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
CHEBI:63005
[Na+].[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Irritating to eyes, respiratory system and skin. May cause cancer. Wear personal protective equipment. Keep away from clothing and other combustible materials. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:1
Instability:3

DOTInformation : DOT Class 5.1, : Oxidizer

SDS Documents

Documents

Promotions

Promotions