missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium nitrate, Analytical Standard, ≥99.995% trace metals basis, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 7631-99-4 |
| Physical Form | Neat |
| Linear Formula | NaNO3 |
| MDL Number | MFCD00011119 |
| Melting Point | 306°C (decomposition) |
| Synonym | E251; Chile salpeter |
| Solubility | Soluble in water |
| Quantity | 1 g |
| Formula Weight | 84.99 |
| Percent Purity | ≥99.995% (trace metals basis) |
| Show More |